Learn More
Medchemexpress LLC Calcineurin autoinhibitory peptide | 148067-21-4 | 100.0% | 2930.32 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP124710MG
Encompass_Preferred
Calcineurin autoinhibitory peptide is a selective inhibitor of Ca2+/calmodulin-dependent protein phosphatase (calcineurin). It could protect neurons from excitatory neuronal death.
  • Selective inhibitor of calcineurin
  • IC50 of approximately 10 μM
  • Protects neurons from excitatory neuronal death
Catalog No. 50-004-22152