Learn More
Cayman Chemical CalcInurInAaIsoformhuma50ug
SDP
Supplier:  Cayman Chemical 3463050 UG
Encompass_Preferred
Source: Active recombinant human N-terminal His-tagged calcineurin A expressed in insect cells • Amino acids: 2-521 (full length) • MW: 60.8 kDa
Catalog No. 50-294-8044