Learn More
Selleck Chemical LLC Calcipotriene 5mg 112965-21-6 Calcipotriol, MC903
SDP
Supplier:  Selleck Chemical LLC S37395MG
Encompass_Preferred
Calcipotriene (Calcipotriol, MC903) is a synthetic derivative of calcitriol, a form of vitamin D. It can induce differentiation and suppresses proliferation of keratinocytes, reversing abnormal keratinocyte changes in psoriasis, and lead to normalization of epidermal growth. Calcipotriol is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8368