Learn More
Apexbio Technology LLC Calcipotriol, 112965-21-6, MFCD00866630, 10 mg
SDP
Supplier:  Apexbio Technology LLC A327110
Encompass_Preferred
MF: C27H40O3, MW: 412.60, Synonyms: MC 903; Calcipotriene. Solubility: 16.7 mg/mL in DMSO, 87.6 mg/mL in EtOH with gentle warming, insoluble in H2O. Calcipotriol is an analogue of vitamin D3.
Catalog No. 50-115-0537