Learn More
TARGETMOL CHEMICALS INC CALCIPOTRIOL 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1485625MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Calcipotriol (MC 903) is a synthetic VitD3 analogue. Which has a high affinity for the vitamin D receptor. purity: 99%
Catalog No. 50-257-7375