Learn More
Medchemexpress LLC HY-10001 50mg , Calcipotriol CAS:112965-21-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000150MG
Encompass_Preferred
HY-10001 50mg Calcipotriol CAS: 112965-21-6 Calcipotriol is a synthetic VitD3 analogue with a high affinity for the vitamin D receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9411