Learn More
Cayman Chemical Calcipotriol (hydrate), 112828-00-9, 10 mg
SDP
Supplier:  Cayman Chemical 1000959910
Encompass_Preferred
Molecular Formula C27H40O3 • H2O, Purity ≥98%, Formula Weight 430.6, Synonyms: 24-cyclopropyl-(1α,3β,5Z,7E,22E,24S)-9,10-secochola-5,7,10(19),22-tetraene-1,3,24-triol, monohydrate, Calcipotriene; A low-calcemic vitamin D receptor agonist; increases keratinocyte differentiation and reduces expression of transcriptional activators in the inflammatory response; induces T-cell differentiation in keratinocytes.
Catalog No. NC1022293
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.