Learn More
Apexbio Technology LLC Calcipotriol monohydrate 147657-22-5 50mg
SDP
Supplier:  Apexbio Technology LLC A327250
Encompass_Preferred
Calcipotriol monohydrate (CAS 147657-22-5) is a synthetic analog of vitamin D3 designed to modulate keratinocyte activity It acts by binding to the vitamin D receptor leading to inhibition of keratinocyte proliferation and induction of their physiological differentiation These effects collectively reduce abnormal epidermal growth and the formation of scaling plaques Calcipotriol monohydrate is widely utilized in biomedical research to investigate the regulation of epidermal differentiation vitamin D receptor-mediated signaling and the pathophysiology of disorders such as psoriasis
Catalog No. 50-000-94940