Learn More
CPC Scientific H-Cys-Ala-Ser-Leu-Ser-Thr-Cys-Val-Leu-Gly-Lys-Leu-Ser-Gln-Glu-Leu-His-Lys-Leu-Gln-Thr-Tyr-Pro-Arg-Thr-Asp-Val-Gly-Ala-Gly-Thr-Pro-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110027
Encompass
Single chain peptide with 32 amino acids that stimulates bone formation by inhibiting osteoblasts, induces bone reabsorption and increases cAMP levels in osteoclasts.ONE-LETTER SEQUENCE: CASLSTCVLGKLSQELHKLQTYPRTDVGAGTP-NH2 (Cys1 and 7 bridge)MOLECULAR FORMULA: C145H240N42O46S2MOLECULAR WEIGHT:3371.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [100016-62-4], RESEARCH AREA: OsteoporosisREFERENCES: T. Kurihara et al., Peptide Chemistry, 1985, 173 (1986)
Catalog No. 50-211-0027 Supplier CPC Scientific Supplier No. OSTP016A
Single chain peptide with 32 amino acids that stimulates bone formation by inhibiting osteoblasts, induces bone reabsorption and increases cAMP levels in osteoclasts.ONE-LETTER SEQUENCE: CASLSTCVLGKLSQELHKLQTYPRTDVGAGTP-NH2 (Cys1 and 7 bridge)MOLECULAR FORMULA: C145H240N42O46S2MOLECULAR WEIGHT:3371.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [100016-62-4], RESEARCH AREA: OsteoporosisREFERENCES: T. Kurihara et al., Peptide Chemistry, 1985, 173 (1986)