Learn More
Medchemexpress LLC HY-P2273 5mg Medchemexpress, Calcitonin (human) CAS:21215-62-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP22735MG
Encompass_Preferred
Medchemexpress, HY-P2273 5mg Calcitonin (human) CAS:21215-62-3 Calcitonin human is an endogenous calcitonin receptor agonist.Calcitonin human lowers systemic blood calcium levels and inhibits bone resorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3082