Learn More
Enzo Life Sciences Calcitonin (salmon) (5mg). CAS: 47931-85-1
SDP
Supplier:  Enzo Life Sciences BMLP4200005
Encompass_Preferred
Calcium-regulating peptide hormone released from the thyroid. Lowers calcium concentration in the blood and decreases bone resorption by osteoclasts. Used for therapeutic applications in osteoporosis. Salmon calcitonin peptide sequence differs from mammalian but is more potent. Purity: ≥98% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water (1mg/ml) or 0.05M acetic acid. Long Term Storage: -20°C.
Catalog No. 50-201-1349