Learn More
Medchemexpress LLC Calcium 2-hydroxypropanoate pentahydrate | 5743-47-5 | 99.8% | 500 G
SDP
Supplier:  Medchemexpress LLC HYB2227D500G
Encompass_Preferred
Calcium 2-hydroxypropanoate (pentahydrate) is a hydroxycarboxylic acid receptor 1 (HCAR1) activator and an epigenetic modulator inducing lysine residues lactylation. It is a glycolysis end-product, bridging the gap between glycolysis and oxidative phosphorylation. Calcium 2-hydroxypropanoate (pentahydrate) is also an oncometabolite and plays an immune protective role of lactate in anti-tumor immunity.
  • HCAR1 Activator
  • Epigenetic modulator inducing lysine residues lactylation
  • Glycolysis end-product
  • Oncometabolite with an immune protective role of lactate in anti-tumor immunity
Catalog No. 50-004-10418