Learn More
Medchemexpress LLC Calcium carbonate | 471-34-1 | MFCD00010906 | 99.2% | 100.09 g/mol | CaCO3 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW243408C100G
Encompass_Preferred
Calcium carbonate (CaCO3) is an inorganic reagent commonly used as a thickener, acidity regulator, and source of calcium in laboratory research. The material is supplied as a fine powder with high purity and is accompanied by a certificate of analysis and a product datasheet to support quality control and handling.
  • High purity (≈99.2%) suitable for research applications.
  • Insoluble in water and soluble in dilute acids.
  • Provided with certificate of analysis and product datasheet for traceability.
  • Available in multiple pack sizes for flexible laboratory use.
  • Commonly used as a buffer, filler, and general reagent in life science workflows.
Catalog No. 50-002-83219