Learn More
Medchemexpress LLC Calcium carbonate | 471-34-1 | MFCD00010906 | 99.2% | 100.09 g/mol | CaCO3 | 500 G

Supplier: Medchemexpress LLC HYW243408C500G
Calcium carbonate (CaCO3) is an inorganic salt provided as a white to off-white solid for laboratory and formulation use. The product is listed at 99.16% purity, has a molecular weight of 100.09 g/mol, and CAS number 471-34-1. Store at room temperature and keep dry and cool.
- High purity for research applications.
- White to off-white solid form for easy handling.
- Chemical formula CaCO3 with molecular weight 100.09 g/mol.
- Supplied in a convenient 500 G pack size for laboratory use.
- Store at room temperature and keep dry and cool.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.