Learn More
Medchemexpress LLC Calcium channel-modulator-1 | 136941-70-3 | 579.45 | 1 ML

Supplier: Medchemexpress LLC HYU0013510MM/1ML
Calcium channel-modulator-1 is an orally active calcium channel modulator that blocks aortic contraction with an IC50 of 0.8 μM. It can be used for the study of cardiovascular conditions.
- Orally active calcium channel modulator
- Blocks aortic contraction with an IC50 of 0.8 μM
- Used for the study of cardiovascular conditions
- Purity: 98.77%
- Molecular weight: 579.45
- CAS number: 136941-70-3
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.