Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride dihydrate | 10035-04-8 | MFCD00149613 | 500g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C3881500G
Encompass
Calcium chloride dihydrate | Purity: ≥99.0% | MW: 147.01 | 10035-04-8 | MFCD00149613 | 500g
Catalog No. NC1747390