Learn More
Medchemexpress LLC Calcium chloride anhydrous | 10043-52-4 | MFCD00010903 | 97.0% | 110.98 g/mol | CaCl2 | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0406E100MG
Encompass_Preferred
Calcium chloride anhydrous for cell culture is an inorganic salt for the preparation of various buffers[1] Calcium chloride anhydrous for cell culture can be used for animal modeling to construct abdominal aortic aneurysm models[3]
Catalog No. 50-002-68493