Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride anhydrous500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C4901500G
Encompass
Calcium chloride (CaCl2) is inexpensive easily available calcium salt with a wide range of applications. It is a key component in deicers as it melts snow and ice. Reports suggest that CaCl2 helps in preserving the firmness of fresh-cut fruits and vegetables during the storage.
Catalog No. 50-289-8368