Learn More
FUJIFILM BIOSCIENCES INC Calcium Chloride Dihydrate, JP grade, 500 g
Supplier:  FUJIFILM BIOSCIENCES INC 03019555
Encompass_Preferred
Pharmaceutical-grade calcium chloride dihydrate (CertiPro: JP) supplied as a white crystalline solid for manufacturing and laboratory applications. It offers consistent assay and lot-to-lot quality for production and analytical workflows.
  • Meets Japanese Pharmacopoeia quality standards.
  • High assay purity, 96.7-103.3% by titration.
  • Molecular formula CaCl2·2H2O, molecular weight 147.01.
  • Packaged in a 500 g container suitable for production use.
  • Consistent lot-to-lot quality for reproducible results.
Catalog No. NC3953036