Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium chloride dihydrate500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 2235066X500G
Encompass
Calcium chloride dihydrate is a moisture resistant cheap and commonly available calcium salt. Its efficacy as a chiral catalyst for various asymmetric organic reactions has been evaluated.
Catalog No. 50-289-9767