Learn More
Medchemexpress LLC Calcium dobesilate (Standard) | 20123-80-2 | 99.8% | 418.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY111603R50MG
Encompass_Preferred
Calcium dobesilate (Standard) is the analytical standard of Calcium dobesilate. It is intended for research and analytical applications.
  • Used as a reference standard for assay.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Vasoprotective agent widely used in chronic venous disease, diabetic retinopathy, and the symptoms of haemorrhoidal attack.
  • For research use only.
Catalog No. 50-004-39676