Learn More
Medchemexpress LLC Calcium fluoride | 7789-75-5 | MFCD00010907 | ≥99.0% | 78.07 | CaF2 | 1 KG

Supplier: Medchemexpress LLC HYW0199621KG
Calcium fluoride (CaF2) is an inorganic salt supplied as an anhydrous white powder for research use. It is used as a reagent and biochemical assay material in laboratory and analytical applications. Product documentation, including a datasheet and safety data sheet, is available.
- Anhydrous white powder form for stable storage.
- High purity suitable for biochemical and analytical applications (≥99.0%).
- Molecular weight 78.07 and chemical formula CaF2 for unambiguous identification.
- Available in multiple pack sizes, including 1 kg, for bulk use.
- Product documentation available: datasheet and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.