Learn More
Strem, An Ascensus Company CAS# 7789-78-8. 50g. Calcium hydride, 95%. MFCD00010897
SDP
Supplier:  Strem, An Ascensus Company 93202150G
Encompass
CAS# 7789-78-8. 50g. Calcium hydride, 95%. MFCD00010897. Molecular Weight: 42.10. Molecular Formula: CaH2. Color/form: 1-20mm light gray granules Strem# 93-2021. http://www.strem.com/catalog/v/93-2021/
Catalog No. 50-901-14355