Learn More
Medchemexpress LLC Calcium hydrogen phosphate dihydrate | 7789-77-7 | MFCD00149621 | 99.0% | 172.09 | CaH5O6P | 1kg
SDP
Supplier:  Medchemexpress LLC HYY07081KG
Encompass_Preferred
Calcium hydrogen phosphate dihydrate (Calcium phosphate dihydrate) can be found quite frequently in urinary calculi stones Calcium hydrogen phosphate dihydrate can be used as an excipient such as diluent adsorbent Pharmaceutical excipients or pharmaceutical auxiliaries refer to other chemical substances used in the pharmaceutical process other than pharmaceutical ingredients Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations which can improve the stability solubility and processability of pharmaceutical preparations Pharmaceutical excipients also affect the absorption distribution metabolism and elimination (ADME) processes of co-administered drugs[1][2]
Catalog No. 50-002-67812