Learn More
Medchemexpress LLC Calcium hydroxide | 1305-62-0 | 95% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYB2217D
Encompass_Preferred
Calcium hydroxide, ACS, 95% possesses high bactericidal activity and high pH. It has the ability to induce hard tissue formation and can dissociate into calcium (Ca) and hydroxyl ions (OH). This product shows potential for intra-canal medicament research.
  • High bactericidal activity
  • High pH
  • Ability to induce hard tissue formation
  • Can dissociate into calcium (Ca) and hydroxyl ions (OH)
  • Potential for intra-canal medicament research
Catalog No. 50-004-14328