Learn More
Chem-Impex International, Inc. Calcium hydroxide, 1305-62-0, MFCD00010901, 1KG
Supplier:  Chem-Impex International, Inc. 012291KG
Encompass_Preferred
Molecular Formula Ca(OH)2; Purity Limit 95.0% (Titration); Grade ACS reagent; Molecular Weight 74.09; Absorbant for carbon dioxide.
Catalog No. 50-492-808