Learn More
Cayman Chemical Calcium Influx Inducer Compou
SDP
Supplier:  Cayman Chemical 419861 MG
Encompass_Preferred
An inducer of calcium influx; enhances EV release from mBMDCs at 10 µM; increases surface levels of CD86 and CD80 on mBMDCs at 10 µM, an effect that can be blocked by BTP2, and increases CD86 and CD80 levels on mBMDC-derived EVs
Catalog No. 50-294-3920