Learn More
Medchemexpress LLC Calcium L-Threonate | 70753-61-6 | 98.0% | 156.14 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0092081G
Encompass_Preferred
Calcium L-Threonate is a calcium aldonate and a calcium supplement. It can be used for the study of osteoporosis. This compound inhibits the bone resorption of osteoclasts in vitro and stimulates ascorbic acid uptake in human T-lymphoma cells.
  • Calcium aldonate
  • Calcium supplement
  • Used for the study of osteoporosis
  • Inhibits bone resorption of osteoclasts in vitro
  • Stimulates ascorbic acid uptake in human T-lymphoma cells
Catalog No. 50-004-53153