Learn More
Medchemexpress LLC Calcium L-Threonate | 70753-61-6 | 98.0% | 156.14 | 500 MG

Supplier: Medchemexpress LLC HYW009208500MG
Calcium L-Threonate is a calcium aldonate and a calcium supplement. It is intended for research purposes only, specifically for studies related to osteoporosis. This product inhibits the bone resorption of osteoclasts in vitro and has been observed to stimulate ascorbic acid uptake in human T-lymphoma cells in a dose-dependent manner.
- Solid, white to off-white appearance.
- Classified as ketones, aldehydes, and acids.
- Derived from endogenous metabolite.
- Ships at room temperature in continental US.
- Store at 4°C, sealed, away from moisture.
- When in solvent, store at -80°C for 6 months or -20°C for 1 month (sealed, away from moisture).
- Soluble in H2O at 5 mg/mL (32.02 mM) with ultrasonic and warming to 60°C.
- Inhibits bone resorption of osteoclasts.
- Stimulates ascorbic acid uptake in human T-lymphoma cells.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.