Learn More
Medchemexpress LLC Calcium N5-methyltetrahydrofolate | 26560-38-3 | 99.9% | 497.52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY175575MG
Encompass_Preferred
Calcium N5-methyltetrahydrofolate is a folic acid supplement that serves as a derivative of Levomefolic acid. It is used in research for conditions such as neural tube defects, infertility, non-Hodgkin's lymphoma, and endometriosis. This compound effectively bypasses folate insufficiency due to MTHFR deficiency in cells with low MTHFR activity.
  • Bypasses folate insufficiency
  • Suitable for research on neural tube defects
  • Applicable for infertility studies
  • Used in research for non-Hodgkin's lymphoma and endometriosis
Catalog No. 50-003-86438