Learn More
Medchemexpress LLC Calcium N5-methyltetrahydrofolate | 26560-38-3 | 99.9% | 497.52 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1755750MG
Encompass_Preferred
Calcium N5-methyltetrahydrofolate, a Levomefolic acid derivative, is a folic acid supplement. It can be used for the research of neural tube defects, infertility, non-hodgkin's lymphoma, and endometriosis.
  • Effectively bypasses folate insufficiency due to MTHFR deficiency in cells with low MTHFR activity.
  • Uptake by human umbilical vascular endothelial cells is inhibited by folic acid.
Catalog No. 50-003-86371