Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium pantothenate meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10) | 137-08-6 | MFCD00002766 | 1KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C87311KG
Encompass
Calcium pantothenate meets USP testing specifications, monograph mol wt. 476.53 (C18H32CaN2O10) | Mol Wt: 238.27 | 137-08-6 | MFCD00002766 | 1KG
Catalog No. 50-185-5520