Learn More
Medchemexpress LLC Calcium phosphate | 7757-93-9 | 98.2% | 500 G

Supplier: Medchemexpress LLC HYB2242500G
Calcium phosphate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
- Solid, white to off-white appearance.
- Molecular weight of 136.06.
- Chemical formula CaHO4P.
- Soluble in H2O at 1.94 mg/mL (14.26 mM) under specific conditions.
- Recommended storage at room temperature for 3 years, or in solvent at -80°C for 2 years, -20°C for 1 year.
- Ships at room temperature in continental US.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.