Learn More
Strem, An Ascensus Company CAS# 10031-30-8. 250g. Calcium phosphate, monobasic, monohydrate, 99%. MFCD00149602
SDP
Supplier:  Strem, An Ascensus Company 202037250G
Encompass
CAS# 10031-30-8. 250g. Calcium phosphate, monobasic, monohydrate, 99%. MFCD00149602. Molecular Weight: 234.06 (252.07). Molecular Formula: Ca(H2PO4)2∙H2O. Color/form: white pwdr. Strem# 20-2037. http://www.strem.com/catalog/v/20-2037/
Catalog No. 50-901-10680