Learn More
Medchemexpress LLC Calcium phosphorylcholine-d9 chloride | 344299-43-0 | 99.85% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB2233AS1MG
Encompass_Preferred
Calcium phosphorylcholine-d9 (chloride) is the deuterium-labeled Calcium phosphorylcholine chloride. It is the main phospholipid component in eukaryotic biofilms and exists in commensal or pathogenic bacteria associated with eukaryotes in prokaryotes, exhibiting a range of immunomodulatory properties.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-10557