Learn More
Medchemexpress LLC Calcium propionate | 4075-81-4 | 99.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB1580100G
Encompass_Preferred
Calcium propionate is an orally active auxiliary agent that helps prevent milk fever and improve the health of dairy cows. It is suitable for research applications related to dairy cow milk fever and subclinical hypocalcemia.
  • Orally active auxiliary agent
  • Prevents milk fever in dairy cows
  • Improves dairy cow health
  • Suitable for research on dairy cow milk fever
  • Suitable for research on subclinical hypocalcemia
Catalog No. 50-004-12490