Learn More
Strem, An Ascensus Company CAS# 7790-76-3. 25g. Calcium pyrophosphate (99.95%-Ca). MFCD00015983
SDP
Supplier:  Strem, An Ascensus Company 93207325G
Encompass
CAS# 7790-76-3. 25g. Calcium pyrophosphate (99.95%-Ca). MFCD00015983. Molecular Weight: 254.10. Molecular Formula: Ca2P2O7. Color/form: 6-8 micron white pwdr. Strem# 93-2073. http://www.strem.com/catalog/v/93-2073/
Catalog No. 50-901-14382