Learn More
Medchemexpress LLC Calcium-sensing R/Ca 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7940810UG
Encompass_Preferred
Calcium-sensing receptor (CaSR) is a G-protein-coupled receptor that is essential for maintaining calcium homeostasis It senses changes in extracellular calcium concentration regulates parathyroid hormone (PTH) production and activates the phosphatidylinositol-calcium second messenger system Calcium-sensing R/CaSR Protein Human (HEK293 His) is the recombinant human-derived Calcium-sensing R/CaSR protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-94755