Learn More
Sigma Aldrich Fine Chemicals Biosciences Calcium Standard for AAS | 250mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 69349250ML
Encompass
TraceCERT, 1000 mg/L Ca in 2% nitric acid, prepared with high purity CaCO3, HNO3 and water.
Catalog No. NC1598567