Learn More
Strem, An Ascensus Company CAS# 1592-23-0. 100g. Calcium stearate, min. 85%. MFCD00036390
SDP
Supplier:  Strem, An Ascensus Company 932045100G
Encompass
CAS# 1592-23-0. 100g. Calcium stearate, min. 85%. MFCD00036390. Molecular Weight: 607.04. Molecular Formula: Ca(O2C18H35)2. Color/form: white pwdr. Strem# 93-2045. http://www.strem.com/catalog/v/93-2045/
Catalog No. 50-901-14374