Learn More
Medchemexpress LLC Calcium trinatrium diethylenetriaminepentaacetic acid hydrate | 207226-35-5 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HY1283701G
Encompass_Preferred
Calcium trinatrium diethylenetriaminepentaacetic acid hydrate is a metal chelator and a useful antidote (e.g., acute cadmium intoxication). It is also a nontoxic inhibitor of CMV replication.
  • Metal chelator
  • Useful antidote for acute cadmium intoxication
  • Nontoxic inhibitor of CMV replication
  • Synonym: Ca-DTPA trisodium salt hydrate
  • Targets CMV
  • Anti-infection pathway
  • Antiviral activity suppressed by metal ions
  • Inhibits CMV L-antigen production with EC50 ranging from 6.1 to 9.9 μM
Catalog No. 50-003-38919