Learn More
TARGETMOL CHEMICALS INC Caldaret HCl 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T843681MG
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Caldaret HCl is an intracellular Ca2+ modulator that regulates calcium homeostasis at the sarcoplasmic reticulum and cell membrane, and can be used to study acute myocardial infarction and heart failure. Purity 99.06%
Catalog No. 50-287-9158