Learn More
Genetex Inc Caldesmon Monoclonal Antibody [MSVA-538R] HistoMAX™ (GTX04451) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04451
Encompass_Preferred
The Caldesmon antibody [MSVA-538R] HistoMAX™ (GTX04451) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human uterine leiomyosarcoma tissue using GTX04451 Caldesmon antibody [MSVA-538R] HistoMAX. Leiomyosarcoma showing intense caldesmon immunostaining of tumor cells Caldesmon immunohistochemistry.
Catalog No. NC3667724