Learn More
Cayman Chemical CaldesmonCTRm RabbIt Mon 100uL
SDP
Supplier:  Cayman Chemical 32318100 UL
Encompass_Preferred
A σ1 receptor ligand (Ki 1.7 nM) selective for σ1 over σ2 receptors (Ki 25.2 nM) inhibits the growth and migration of PC3 prostate cancer and A549 NSCLC cells as well as the migration of C32 melanoma and U373 MG glioblastoma cells at 10 UM increases survival in a U373 MG mouse xenograft model at 2 mg/kg increases firing of serotonergic neurons in the rat dorsal raphe nucleus at 2 mg/kg
Catalog No. 50-294-8482