Learn More
Medchemexpress LLC HY-N6850 5mg Medchemexpress, Calenduloside E CAS:26020-14-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN68505MG
Encompass_Preferred
Medchemexpress, HY-N6850 5mg Calenduloside E CAS:26020-14-4 Calenduloside E (CE) is a natural pentacyclic triterpenoid saponin extracted from Aralia elata. Calenduloside E (CE) has anti-apoptotic potent by targeting heat shock protein 90 (Hsp90). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0476