Learn More
Qc Supply LLC CALF DRENCHER 2.5L
SDP
Supplier:  Qc Supply LLC 140628
Encompass
Calf Drencher from JorVet™ has a collapsible 2 1/2 liter drench bag with special probe. Rigid top portion with holes for easy handling or hanging and comes with on/off flow clamp. The special cold temperature-resistant plastic will not crack and is graduated with markings in both liters and quarts. Easy filling and a round ball-shape end to aid in proper placement.Instructions — Fill with desired fluid for administration. Pass esophageal tube into the mouth and down the throat angling for the left back corner of the mouth for proper placement down the esophagus. Once the tube is in place, release the clamp to allow fluid to flow while holding bag above the animal’s head. Once fluid has run in, clamp the tube off and slowly and gently remove from throat to avoid injury.
Catalog No. NC2393711