Learn More
Aat Bioquest Caliber Fluor™ 580
SDP
Supplier:  Aat Bioquest 95
Encompass_Preferred
Caliber Fluor™ 580 is an extremely stable fluorophore that has its absorption and fluorescence independent of pH change from pH 3 to 10.
Catalog No. 50-297-9294