Learn More
Sigma Aldrich Fine Chemicals Biosciences Color Reference Solutions | 7 x 60 ML Bottles
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 9254701SET
Encompass
Color Reference Solutions | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | 7 x 60 mL bottlesAnalytical standards composed of stable liquid color solutions used for calibration and measurement of color intensity across the brown, yellow, orange and red color spectrum. Applicable for precise colorimetric calibration of colorimeters and spectrophotometers.
Catalog No. NC2373534