Learn More
Andwin Scientific CALIBRATING BUFFER PH 4.0 15ML
SDP
Supplier:  Andwin Scientific 2283436
Encompass
The buffer solution is a high quality buffer ideal for pH and conductivity. This solution is measured to have a pH of 4.01 0.02 at 25C. It is packaged in a 15 mL bottle.
Catalog No. NC9972510