Learn More
Electron Microscopy Sciences Calibration Specimen 150-1D, Steel disk for AFM
SDP
Catalog No. 501901660
Encompass_Preferred
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Has a nominal period (pitch) of 150 nm, one dimensional, fabricated on a transparent substrate (Aluminum lines on glass).
Mount: AFM
Catalog No. 50-190-1660 Supplier Electron Microscopy Sciences Supplier No. 801251DAFM
Add to Cart
Add to Cart
SPM calibration specimens are produced by holographic fabrication that assures high accuracy and precision. The standards provide easier testing of your SPM, improved accuracy of critical dimension measurements and accuracy of 0.5% (1 std. dev.). The pattern height of greater than 100nm provides excellent image contrast. There is uniform coverage of entire chip which allows for imaging anywhere on the chip. Has a nominal period (pitch) of 150 nm, one dimensional, fabricated on a transparent substrate (Aluminum lines on glass).
Mount: AFM